726753
(S)-(−)-1-Phenylethanol
ChiPros®, produced by BASF, 98%
Synonym(s):
(−)-Methyl phenyl carbinol, (S)-(−)-α-Methylbenzyl alcohol, (S)-(−)-sec-Phenylethyl alcohol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
grade
produced by BASF
Assay
98%
optical purity
ee: ≥97.5%
bp
88-89 °C/10 mmHg (lit.)
mp
9-11 °C (lit.)
density
1.012 g/mL at 20 °C (lit.)
SMILES string
C[C@H](O)c1ccccc1
InChI
1S/C8H10O/c1-7(9)8-5-3-2-4-6-8/h2-7,9H,1H3/t7-/m0/s1
InChI key
WAPNOHKVXSQRPX-ZETCQYMHSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
ChiPros is a registered trademark of BASF SE
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2
WGK
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Scientific reports, 7(1), 3081-3081 (2017-06-10)
(S)-carbonyl reductase II (SCRII) from Candida parapsilosis is a short-chain alcohol dehydrogenase/reductase. It catalyses the conversion of 2-hydroxyacetophenone to (S)-1-phenyl-1,2-ethanediol with low efficiency. Sortase was reported as a molecular "stapler" for site-specific protein conjugation to strengthen or add protein functionality.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service