![(S)-(−)-1-Phenylethanol ≥98.5% (sum of enantiomers, GC)](/deepweb/assets/sigmaaldrich/product/structures/315/238/1cd53251-231f-4b33-92dc-73321e6639eb/640/1cd53251-231f-4b33-92dc-73321e6639eb.png)
(S)-(−)-1-Phenylethanol
![(S)-(−)-1-Phenylethanol ≥98.5% (sum of enantiomers, GC)](/deepweb/assets/sigmaaldrich/product/structures/315/238/1cd53251-231f-4b33-92dc-73321e6639eb/640/1cd53251-231f-4b33-92dc-73321e6639eb.png)
Synonym(s):
(−)-Methyl phenyl carbinol, (S)-(−)-α-Methylbenzyl alcohol, (S)-(−)-sec-Phenylethyl alcohol
Empirical Formula (Hill Notation):
C8H10O
CAS Number:
Molecular Weight:
122.16
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing