Skip to Content
Merck
All Photos(1)

Key Documents

469653

Sigma-Aldrich

(R)-(+)-Mandelonitrile

97%

Synonym(s):

(R)-(+)-α-Hydroxybenzeneacetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OH)CN
CAS Number:
Molecular Weight:
133.15
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

Assay

97%

optical activity

[α]20/D +42°, c = 1 in chloroform

refractive index

n20/D 1.532 (lit.)

bp

170 °C (lit.)

mp

28-30 °C (lit.)

density

1.117 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

O[C@@H](C#N)c1ccccc1

InChI

1S/C8H7NO/c9-6-8(10)7-4-2-1-3-5-7/h1-5,8,10H/t8-/m0/s1

InChI key

NNICRUQPODTGRU-QMMMGPOBSA-N

Application

Useful intermediate in the preparation of optically active α-hydroxy carboxylic acids, α-hydroxy aldehydes, α-hydroxy ketones, and 2-amino alcohols.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daisuke Matsui et al.
Scientific reports, 7(1), 9558-9558 (2017-08-27)
Insolubility of proteins expressed in the Escherichia coli expression system hinders the progress of both basic and applied research. Insoluble proteins contain residues that decrease their solubility (aggregation hotspots). Mutating these hotspots to optimal amino acids is expected to improve
Raimondas Mozuraitis et al.
Scientific reports, 9(1), 14262-14262 (2019-10-05)
The male butterfly Pieris napi produces the anti-aphrodisiac pheromone methyl salicylate (MeS) and transfers it to the female during mating. After mating she releases MeS, when courted by conspecific males, which decreases her attractiveness and the duration of male harassment
Kruse, C.G. In Collins, A.N. Sheldrake, G.N. Crosby, J., Eds.
Chirality in Industry, 279-279 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service