Skip to Content
Merck
All Photos(1)

Key Documents

442542

Sigma-Aldrich

Methylmercury(II) iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3HgI
CAS Number:
Molecular Weight:
342.53
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

reaction suitability

reagent type: catalyst
core: mercury

mp

145 °C (lit.)

SMILES string

C[Hg]I

InChI

1S/CH3.Hg.HI/h1H3;;1H/q;+1;/p-1

InChI key

JVDIOYBHEYUIBM-UHFFFAOYSA-M

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S C Bondy et al.
Toxicology letters, 58(1), 13-21 (1991-09-01)
The effect of a neurotoxic organo-metal, methyl mercuric iodide, and an aromatic solvent, toluene, upon the transmembrane potential (psi), across both the limiting membrane of isolated nerve terminals and their mitochondria, has been studied. Exposure of nerve endings to either
K Okamura-Ikeda et al.
The Journal of biological chemistry, 260(2), 1338-1345 (1985-01-25)
Medium-chain and long-chain acyl-CoA dehydrogenases from rat liver have been purified in two forms, holoenzymes containing FAD and apoenzymes which do not contain this cofactor. In contrast, short-chain acyl-CoA dehydrogenase can only be isolated as the holoenzyme. Marked differences in
[Effect of methylmercury iodide on the rate of regeneration of epithelial crypts of the rat large intestine].
L A Khvylia et al.
Voprosy pitaniia, (3)(3), 73-74 (1984-05-01)
M S Benjumovich
Toxicology in vitro : an international journal published in association with BIBRA, 15(6), 671-675 (2001-11-08)
The median lethal cell culture dose (CCLD(50)) and its 95% confidence interval were determined using the Behrens and Kärber area method with subsequent regression analysis by the action of methylmercury iodide (CH(3)HgI; MMI) on monolayer cultures of HeLa cells. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service