Merck
  • An essential cysteine residue located in the vicinity of the FAD-binding site in short-chain, medium-chain, and long-chain acyl-CoA dehydrogenases from rat liver mitochondria.
An essential cysteine residue located in the vicinity of the FAD-binding site in short-chain, medium-chain, and long-chain acyl-CoA dehydrogenases from rat liver mitochondria.
The Journal of biological chemistry (1985-01-25)
K Okamura-Ikeda, Y Ikeda, K Tanaka
ABSTRACT
Medium-chain and long-chain acyl-CoA dehydrogenases from rat liver have been purified in two forms, holoenzymes containing FAD and apoenzymes which do not contain this cofactor. In contrast, short-chain acyl-CoA dehydrogenase can only be isolated as the holoenzyme. Marked differences in the reactivity to organic sulfhydryl reagents were observed between the apo and holo forms of these enzymes. While the two apoenzymes were severely inactivated by N-ethylmaleimide (NEM), p-chloromercuribenzoate (pCMB), and iodoacetate (IAA), the two corresponding holoenzymes were not susceptible to these reagents. The inactivation of the two apoenzymes by NEM followed pseudo-first order kinetics. Incubation of the apoenzymes with FAD completely prevented the inactivation by the organic sulfhydryl reagents. Methylmercury halides (iodide or chloride) inactivated both the apo and holo forms of medium-chain and long-chain acyl-CoA dehydrogenases. On the other hand, holo-short-chain acyl-CoA dehydrogenase behaved somewhat differently from the other two holoenzymes in that it was inactivated by pCMB (but not NEM or IAA) following a pseudo-first order process. The titration of the two apoenzymes with [14C]NEM and that of the holo-short-chain acyl-CoA dehydrogenase with [14C]pCMB indicated that all three acyl-CoA dehydrogenases contain a single essential cysteine residue/subunit. In the inactivation of holo-medium-chain and holo-long-chain acyl-CoA dehydrogenases with methylmercury halide, the same essential cysteine residue was modified without perturbing or releasing the enzyme-bound FAD. The inactivations of the three holoenzymes by appropriate organic sulfhydryl reagents were prevented by prior incubation with substrate. These experimental results indicate that the essential cysteine residue is located in the vicinity of the FAD- and substrate-binding sites within the active center of the enzymes. It appears, however, that this cysteine residue does not participate directly in FAD binding.