Skip to Content
Merck

Skip To

317543

Phenacyl 4-(bromomethyl)phenylacetate

95%

Synonym(s):

4-(Bromomethyl)phenylacetic acid phenacyl ester

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C17H15BrO3
CAS Number:
Molecular Weight:
347.20
UNSPSC Code:
12352200
NACRES:
NA.22
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
2292032
Assay:
95%


assay

95%

reaction suitability

reagent type: cross-linking reagent

mp

84-87 °C (lit.)

SMILES string

BrCc1ccc(CC(=O)OCC(=O)c2ccccc2)cc1

InChI

1S/C17H15BrO3/c18-11-14-8-6-13(7-9-14)10-17(20)21-12-16(19)15-4-2-1-3-5-15/h1-9H,10-12H2

InChI key

BUQPIKAEKYNDAS-UHFFFAOYSA-N



Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
317543-5G04061833372074

Questions

Reviews

No rating value

Active Filters