Skip to Content
Merck
All Photos(3)

Key Documents

308552

Sigma-Aldrich

5,5-Dibromobarbituric acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H2Br2N2O3
CAS Number:
Molecular Weight:
285.88
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

drug control

USDEA Schedule IIIN; regulated under CDSA - not available from Sigma-Aldrich Canada

mp

240-242 °C (lit.)

functional group

bromo

SMILES string

BrC1(Br)C(=O)NC(=O)NC1=O

InChI

1S/C4H2Br2N2O3/c5-4(6)1(9)7-3(11)8-2(4)10/h(H2,7,8,9,10,11)

InChI key

AMATXUCYHHHHHB-UHFFFAOYSA-N

Application

5,5-Dibromobarbituric acid was used in the synthesis of condensed pteridine system by undergoing condensation with 4,5-diamino-pyrimidines in the presence of pyridine[1].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

THE REACTION OF 5, 5-DIHALOBARBITURIC ACIDS WITH PYRIDINE1, 2.
The Journal of Organic Chemistry, 20(2), 264-270 (1955)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service