Skip to Content
Merck
All Photos(1)

Documents

281727

Sigma-Aldrich

3,3′-(Ethylenedioxy)diphenol

85%, technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4OCH2CH2OC6H4OH
CAS Number:
Molecular Weight:
246.26
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

Assay

85%

form

solid

mp

157-160 °C (lit.)

SMILES string

Oc1cccc(OCCOc2cccc(O)c2)c1

InChI

1S/C14H14O4/c15-11-3-1-5-13(9-11)17-7-8-18-14-6-2-4-12(16)10-14/h1-6,9-10,15-16H,7-8H2

InChI key

DBFHCPVZZSVFJL-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service