C34405
2-Chloro-3′,4′-dihydroxyacetophenone
97%
Synonym(s):
4-(Chloroacetyl)catechol
About This Item
Recommended Products
Quality Level
Assay
97%
form
powder
mp
174-176 °C (lit.)
SMILES string
Oc1ccc(cc1O)C(=O)CCl
InChI
1S/C8H7ClO3/c9-4-8(12)5-1-2-6(10)7(11)3-5/h1-3,10-11H,4H2
InChI key
LWTJEJCZJFZKEL-UHFFFAOYSA-N
Gene Information
human ... PTPN6(5777)
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service