Skip to Content
Merck
All Photos(1)

Key Documents

262382

Sigma-Aldrich

4,4-Dimethyl-3-oxopentanenitrile

99%

Synonym(s):

Pivaloylacetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCOCH2CN
CAS Number:
Molecular Weight:
125.17
Beilstein:
1746674
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

125-126 °C/22 mmHg (lit.)

mp

66-69 °C (lit.)

SMILES string

CC(C)(C)C(=O)CC#N

InChI

1S/C7H11NO/c1-7(2,3)6(9)4-5-8/h4H2,1-3H3

InChI key

MXZMACXOMZKYHJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4,4-Dimethyl-3-oxopentanenitrile has been used in the preparation of:
  • 4,4-dimethyl-3-oxo-2-benzylpentanenitrile[1]
  • 4,4-dimethyl-3-oxo-2-(4-hydroxybenzyl)pentanenitrile[1]
  • 4,4-dimethyl-3-oxo-2-(4-methoxybenzyl)pentanenitrile[1]
  • 4,4-dimethyl-3-oxo-2-(4-fluorobenzyl)pentanenitrile[1]
  • 4,4-dimethyl-3-oxo-2-(3,4-methylenedioxybenzyl)pentanenitrile[1]
  • 1-(5-tert-butyl-2-p-tolyl-2H-pyrazol-3-yl)-3-phenylurea[2]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John Regan et al.
Journal of medicinal chemistry, 45(14), 2994-3008 (2002-06-28)
We report on a series of N-pyrazole, N'-aryl ureas and their mode of binding to p38 mitogen activated protein kinase. Importantly, a key binding domain that is distinct from the adenosine 5'-triphoshate (ATP) binding site is exposed when the conserved
Phillip J Black et al.
Organic & biomolecular chemistry, 4(1), 116-125 (2005-12-17)
Alcohols have been employed as substrates for C-C bond-forming reactions which involve initial activation by the temporary removal of hydrogen to form an aldehyde. The intermediate aldehyde is converted into an alkene via a Horner-Wadsworth-Emmons reaction, nitroaldol and aldol reactions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service