Skip to Content
Merck
All Photos(2)

Documents

D142409

Sigma-Aldrich

2-Chloro-N,N-dimethylpropylamine hydrochloride

98%

Synonym(s):

β-(Dimethylamino)isopropyl chloride hydrochloride, 2-(Dimethylamino)isopropyl chloride hydrochloride, 2-Chloro-1-(dimethylamino)propane hydrochloride, N-(2-Chloropropyl)dimethylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CHClCH2N(CH3)2 · HCl
CAS Number:
Molecular Weight:
158.07
Beilstein:
3670215
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

mp

187-190 °C (lit.)

SMILES string

Cl.CC(Cl)CN(C)C

InChI

1S/C5H12ClN.ClH/c1-5(6)4-7(2)3;/h5H,4H2,1-3H3;1H

InChI key

OCWGRWAYARCRTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service