Skip to Content
Merck
All Photos(1)

Key Documents

245895

Sigma-Aldrich

Methyl nonanoate

98%

Synonym(s):

Methyl pelargonate, Nonanoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7COOCH3
CAS Number:
Molecular Weight:
172.26
Beilstein:
1754521
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

98%

refractive index

n20/D 1.421 (lit.)

bp

213-214 °C (lit.)

density

0.875 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCC(=O)OC

InChI

1S/C10H20O2/c1-3-4-5-6-7-8-9-10(11)12-2/h3-9H2,1-2H3

InChI key

IJXHLVMUNBOGRR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl nonanoate (Methyl pelargonate) was used in the synthesis of vanillyl nonanoate, a model compound of capsinoids[1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenji Kobata et al.
Bioscience, biotechnology, and biochemistry, 66(2), 319-327 (2002-05-10)
Capsinoids are a novel group of compounds produced by the Capsicum plant. We synthesized a capsinoid by the lipase-catalyzed esterification of vanillyl alcohol with fatty acid derivatives in an organic solvent. The use of seven out of 17 commercially available
Cuihua Liu et al.
Journal of the science of food and agriculture, 95(1), 111-119 (2014-04-12)
Terpenoids are major components of carotenoids, limonoids and aromas in citrus fruits, resulting in fruit coloration, bitterness and aroma. In this study the carotenoid, limonoid and volatile profiles of red-flesh Chuhong pummelo (CH) and pale green-flesh Feicui pummelo (FC) were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service