Skip to Content
Merck
All Photos(1)

Key Documents

773948

Sigma-Aldrich

Strontium phosphate

powder, -200 mesh, 99.9% trace metals basis

Synonym(s):

Strontium phosphate tribasic, Tristrontium diphosphate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
CHF 166.00

CHF 166.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
CHF 166.00

About This Item

Linear Formula:
Sr3(PO4)2
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

CHF 166.00


Check Cart for Availability

Request a Bulk Order

Assay

99.9% trace metals basis

form

powder

particle size

-200 mesh

mp

>380 °C

SMILES string

[Sr++].[Sr++].[Sr++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

InChI

1S/2H3O4P.3Sr/c2*1-5(2,3)4;;;/h2*(H3,1,2,3,4);;;/q;;3*+2/p-6

InChI key

JOPDZQBPOWAEHC-UHFFFAOYSA-H

Application

Sr3(PO4)2 is a promising phosphor host material; showing different emission properties upon doping with rare earth and other elements.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poort; S. H. M.
Journal of Solid State Chemistry, 122, 432-432 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service