Skip to Content
Merck
All Photos(1)

Key Documents

230561

Sigma-Aldrich

Sulforhodamine G

Dye content 60 %

Synonym(s):

Acid Red 50

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H25N2NaO7S2
CAS Number:
Molecular Weight:
552.60
Colour Index Number:
45220
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder

composition

Dye content, 60%

λmax

529 nm

SMILES string

[Na+].CCNc1cc2OC3=CC(=N\CC)\C(C)=CC3=C(c2cc1C)c4ccc(cc4S(O)(=O)=O)S([O-])(=O)=O

InChI

1S/C25H26N2O7S2.Na/c1-5-26-20-12-22-18(9-14(20)3)25(19-10-15(4)21(27-6-2)13-23(19)34-22)17-8-7-16(35(28,29)30)11-24(17)36(31,32)33;/h7-13,26H,5-6H2,1-4H3,(H,28,29,30)(H,31,32,33);/q;+1/p-1/b27-21-;

InChI key

NWWFZBYHUXCUDI-DJILMAANSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sulforhodamine G is an ambipolar fluorescent dye that is used as an acceptor dye for organic electronics. It is a water soluble and hydrophilic dye that shows an absorption peak at a wavelength of 529 nm.

Application

Sulforhodamine G can be incorporated in composites as a dye for characterizing fluorescent correlation spectroscopy based images. It can also be used in the development of an acceptor molecule that facilitates the emission of white light in a light emitting hybrid.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Classifying the photophysical dynamics of single-and multiple-chromophoric molecules by single molecule spectroscopy.
Yip W, et al.
The Journal of Physical Chemistry A, 102(39), 7564-7575 (1998)
Fluorescent dye and media properties affecting sorption and tracer selection
Kasnavia, T., Vu, D., & Sabatini, D. A.
Groundwater, 37(3), 376-381 (1999)
Rui Hao et al.
Proceedings of the National Academy of Sciences of the United States of America, 115(23), 5878-5883 (2018-05-23)
Nucleation and growth of hydrogen nanobubbles are key initial steps in electrochemical water splitting. These processes remain largely unexplored due to a lack of proper tools to probe the nanobubble's interfacial structure with sufficient spatial and temporal resolution. We report
An anisotropic ink based on crystalline nanocellulose: Potential applications in security printing.
Chindawong C and Johannsmann D
Journal of Applied Polymer Science, 131(22) (2014)
Highly Pure Solid-State White-Light Emission from Solution-Processable Soft-Hybrids.
Rao KV, et al.
Advanced Materials, 25(12), 1713-1718 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service