Skip to Content
Merck
All Photos(1)

Key Documents

162582

Sigma-Aldrich

4,4′-Dimethoxybenzhydrol

99%

Synonym(s):

Bis(4-methoxyphenyl)carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3OC6H4)2CHOH
CAS Number:
Molecular Weight:
244.29
Beilstein:
748532
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

mp

67-70 °C (lit.)

functional group

hydroxyl

SMILES string

COc1ccc(cc1)C(O)c2ccc(OC)cc2

InChI

1S/C15H16O3/c1-17-13-7-3-11(4-8-13)15(16)12-5-9-14(18-2)10-6-12/h3-10,15-16H,1-2H3

InChI key

ZODAOVNETBTTJX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4,4′-Dimethoxybenzhydrol causes N-alkylation of amides, lactams, carbamates, ureas and anilines in acetic acid during H2SO4 catalysis[1].

Application

4,4′-Dimethoxybenzhydrol was used to study the kinetics and mechanism of oxidation of substituted benzhydrols to corresponding benzophenones in the presence of Hg(OAc)2 by N-bromosuccinimide[2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetics and mechanism of oxidation of some substituted benzhydrols by N-bromosuccinimide.
Hiran BL, et al.
Kinetics and Catalysis, 46(3), 334-339 (2005)
One-Step Hydroxy Substitution of 4, 4'-Dimethoxybenzhydrol with Amides, Lactams, Carbamates, Ureas and Anilines.
Henneuse C, et al.
Synthesis, 1996(04), 495-501 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service