Merck
Glutathione Transferase U13 Functions in Pathogen-Triggered Glucosinolate Metabolism.
Plant physiology (2017-11-11)
Mariola Piślewska-Bednarek, Ryohei Thomas Nakano, Kei Hiruma, Marta Pastorczyk, Andrea Sanchez-Vallet, Suthitar Singkaravanit-Ogawa, Danuta Ciesiołka, Yoshitaka Takano, Antonio Molina, Paul Schulze-Lefert, Paweł Bednarek
ZUSAMMENFASSUNG
Glutathione (GSH) and indole glucosinolates (IGs) exert key functions in the immune system of the model plant Arabidopsis (
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Camalexin, ≥98% (HPLC)