- New chemical and radiochemical routes to [
New chemical and radiochemical routes to [
MedChemComm (2017-12-26)
J A H Inkster, S Zhang, V Akurathi, A Belanger, S Dubey, T Treves, A B Packard
PMID29276578
ZUSAMMENFASSUNG
New chemical and radiochemical syntheses are described for the preparation of [
MATERIALIEN