Merck
  • Clinical and Practical Use of Calcimimetics in Dialysis Patients With Secondary Hyperparathyroidism.
Clinical and Practical Use of Calcimimetics in Dialysis Patients With Secondary Hyperparathyroidism.
Clinical journal of the American Society of Nephrology : CJASN (2015-08-01)
Jordi Bover, Pablo Ureña, César Ruiz-García, Iara daSilva, Patricia Lescano, Jacqueline del Carpio, José Ballarín, Mario Cozzolino
ZUSAMMENFASSUNG
CKD and CKD-related mineral and bone disorders (CKD-MBDs) are associated with high cardiovascular and mortality risks. In randomized clinical trials (RCTs), no single drug intervention has been shown to reduce the high mortality risk in dialysis patients, but several robust secondary analyses point toward important potential beneficial effects of controlling CKD-MBD-related factors and secondary hyperparathyroidism. The advent of cinacalcet, which has a unique mode of action at the calcium-sensing receptor, represented an important step forward in controlling CKD-MBD. In addition, new RCTs have conclusively shown that cinacalcet improves achievement of target levels for all of the metabolic abnormalities associated with CKD-MBD and may also attenuate the progression of vascular and valvular calcifications in dialysis patients. However, a final conclusion on the effect of cinacalcet on hard outcomes remains elusive. Tolerance of cinacalcet is limited by frequent secondary side effects such as nausea, vomiting, hypocalcemia and oversuppression of parathyroid hormone, which may cause some management difficulties, especially for those lacking experience with the drug. Against this background, this review aims to summarize the results of studies on cinacalcet, up to and including the publication of the recent ADVANCE and EVOLVE RCTs, as well as recent post hoc analyses, and to offer practical guidance on how to improve the clinical management of the most frequent adverse events associated with cinacalcet, based on both currently available information and personal experience. In addition, attention is drawn to less common secondary effects of cinacalcet treatment and advisable precautions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cinacalcet hydrochloride, ≥98% (HPLC)