Merck
  • Benzimidazole inhibitors of protein kinase CK2 potently inhibit the activity of atypical protein kinase Rio1.
Benzimidazole inhibitors of protein kinase CK2 potently inhibit the activity of atypical protein kinase Rio1.
Molecular and cellular biochemistry (2016-12-03)
Konrad Kubiński, Maciej Masłyk, Andrzej Orzeszko
ZUSAMMENFASSUNG
Benzimidazole derivatives of 5,6-dichlorobenzimidazole 1-β-D-ribofuranoside (DRB) comprise the important class of protein kinase CK2 inhibitors. Depending on the structure, benzimidazoles inhibit CK2 with different selectivity and potency. Besides CK2, the compounds can inhibit, with similar activity, other classical eukaryotic protein kinases (e.g. PIM, DYRK, and PKD). The present results show that a majority of the most common CK2 inhibitors can affect the atypical kinase Rio1 in a nanomolar range. Kinetic data confirmed the mode of action of benzimidazoles as typical ATP-competitive inhibitors. In contrast to toyocamycin-the first discovered small-molecule inhibitor of Rio1-the most potent representative of benzimidazoles TIBI (IC
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DMAT, ≥98% (HPLC)