Merck
  • Infrared spectroscopic study of absorption and separation of CO using copper(i)-containing ionic liquids.
Infrared spectroscopic study of absorption and separation of CO using copper(i)-containing ionic liquids.
Dalton transactions (Cambridge, England : 2003) (2017-02-09)
Stephen E Repper, Anthony Haynes, Evert J Ditzel, Glenn J Sunley
ZUSAMMENFASSUNG
Absorption of carbon monoxide by copper(i)-containing ionic liquids, [C
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1-Chlorhexan, 99%