Merck
  • Large-scale models of signal propagation in human cells derived from discovery phosphoproteomic data.
Large-scale models of signal propagation in human cells derived from discovery phosphoproteomic data.
Nature communications (2015-09-12)
Camille D A Terfve, Edmund H Wilkes, Pedro Casado, Pedro R Cutillas, Julio Saez-Rodriguez
ZUSAMMENFASSUNG
Mass spectrometry is widely used to probe the proteome and its modifications in an untargeted manner, with unrivalled coverage. Applied to phosphoproteomics, it has tremendous potential to interrogate phospho-signalling and its therapeutic implications. However, this task is complicated by issues of undersampling of the phosphoproteome and challenges stemming from its high-content but low-sample-throughput nature. Hence, methods using such data to reconstruct signalling networks have been limited to restricted data sets and insights (for example, groups of kinases likely to be active in a sample). We propose a new method to handle high-content discovery phosphoproteomics data on perturbation by putting it in the context of kinase/phosphatase-substrate knowledge, from which we derive and train logic models. We show, on a data set obtained through perturbations of cancer cells with small-molecule inhibitors, that this method can study the targets and effects of kinase inhibitors, and reconcile insights obtained from multiple data sets, a common issue with these data.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
AZD5438, ≥97% (HPLC)