Merck
  • [Novel renal function marker, ATP--establishing the normal range, cases of anti-tumor drags administration for urinary-track tumor, diabetic diseases and a newborn baby].
[Novel renal function marker, ATP--establishing the normal range, cases of anti-tumor drags administration for urinary-track tumor, diabetic diseases and a newborn baby].
Rinsho byori. The Japanese journal of clinical pathology (2002-06-25)
Moto-o Nakajima, Yuki Etoh, Mitsuru Yanai, Risako Kawamura, Kazunari Kaneko, Kazumasa Aoyagi, Kenkiti Koiso
ZUSAMMENFASSUNG
Urinary free ATP assay by the firefly luciferin-luciferase method is a rapid and simple method for determining renal function, especially uriniferous tubule function. Normal range of urinary free ATP concentration, daily ATP excretion in urine, urinary ATP/creatinine value and ATP decomposition activity in urine is 1.1 x 10(-9)-3.4 x 10(-8) M, 4.0 x 10(-9)-4.1 x 10(-8) mole, 5.0 x 10(-13)-5.9 x 10(-11) mol/mgCr and 100-77% express for the remaining rate of additional ATP, respectively. A significant correlation was found between free ATP concentration and daily ATP excretion in urine with a correlation coefficient of 0.84. In cases of anti-tumor drug(cisplatin = cis-diamminedichloroplatinum II) administration for urinary-track tumor, abnormal urinary free ATP concentration and ATP decomposition activity in urine were clearly demonstrated after a few days of cisplatin administration. The appearance of a tendency toward abnormal relative ATP values were similar to changes in beta 2-MG and NAG values. Diabetic patients often demonstrate unusually high values of urinary free ATP concentration. In asphyxia of the newborn, urinary ATP/creatinine value were significantly higher than those in healthy newborn, but urinary NAG values did not differ.