Merck
  • β-Siloxy-α-haloketones through highly diastereoselective single and double mukaiyama aldol reactions.
β-Siloxy-α-haloketones through highly diastereoselective single and double mukaiyama aldol reactions.
Chemistry (Weinheim an der Bergstrasse, Germany) (2013-02-21)
Jakub Saadi, Hisashi Yamamoto
ZUSAMMENFASSUNG
Double-action haloketones: A super silyl group enabled the first highly diastereoselective Mukaiyama aldol reactions of α-chloro- and α-fluoroketones with a wide range of aldehydes, providing anti-β-siloxy-α-haloketones. This process is compatible with one-pot double-aldol methodology and allows for rapid access to new halogen-modified polyketide fragments bearing up to four contiguous stereocenters.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3,5-Dibrombenzaldehyd