Merck
  • Novel C-terminally truncated isoforms of the CXC chemokine beta-thromboglobulin and their impact on neutrophil functions.
Novel C-terminally truncated isoforms of the CXC chemokine beta-thromboglobulin and their impact on neutrophil functions.
Journal of immunology (Baltimore, Md. : 1950) (1998-10-30)
J E Ehlert, J Gerdes, H D Flad, E Brandt
ZUSAMMENFASSUNG
The neutrophil agonist neutrophil-activating peptide-2 (NAP-2) arises through proteolytic processing of platelet-derived N-terminally extended inactive precursors, the most abundant one being connective tissue-activating peptide-III (CTAP-III). Apart from N-terminal processing, C-terminal processing also appears to participate in the functional regulation of NAP-2, as indicated by our recent identification of an isoform missing four C-terminal amino acids, NAP-2 (1-66), which was about threefold more potent than full-size NAP-2. In the present study, we report on the discovery and identification of natural NAP-2 (1-63), an isoform truncated by seven C-terminal residues. Functional and receptor-binding analyses demonstrated that NAP-2 (1-63) represents the most active isoform, being about fivefold more potent than full-size NAP-2. Analyses of rNAP-2 isoforms successively truncated at the C terminus by up to eight residues suggest functionally important roles for acidic residues and for the leucine at position 63, a residue highly conserved within CXC chemokines. Finally, we report on a novel C-terminally truncated isoform of CTAP-III (CTAP-III (1-81)) representing the potential precursor of NAP-2 (1-66). We show that C-terminal truncation in CTAP-III enhances its potency to desensitize chemokine-induced neutrophil activation, indicating that C-terminal processing might not only serve to enhance neutrophil activation, but might as well participate in the down-regulation of an inflammatory response.