Merck
MicroRNA-30c targets cytoskeleton genes involved in breast cancer cell invasion.
Breast cancer research and treatment (2012-12-12)
Jessica Bockhorn, Kathy Yee, Ya-Fang Chang, Aleix Prat, Dezheng Huo, Chika Nwachukwu, Rachel Dalton, Simo Huang, Kaitlin E Swanson, Charles M Perou, Olufunmilayo I Olopade, Michael F Clarke, Geoffrey L Greene, Huiping Liu
ZUSAMMENFASSUNG
Metastasis remains a significant challenge in treating cancer. A better understanding of the molecular mechanisms underlying metastasis is needed to develop more effective treatments. Here, we show that human breast tumor biomarker miR-30c regulates invasion by targeting the cytoskeleton network genes encoding twinfilin 1 (TWF1) and vimentin (VIM). Both VIM and TWF1 have been shown to regulate epithelial-to-mesenchymal transition. Similar to TWF1, VIM also regulates F-actin formation, a key component of cellular transition to a more invasive mesenchymal phenotype. To further characterize the role of the TWF1 pathway in breast cancer, we found that IL-11 is an important target of TWF1 that regulates breast cancer cell invasion and STAT3 phosphorylation. The miR-30c-VIM/TWF1 signaling cascade is also associated with clinical outcome in breast cancer patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid