Merck
  • Correlation between B7-H3 expression and rheumatoid arthritis: A new polymorphism haplotype is associated with increased disease risk.
Correlation between B7-H3 expression and rheumatoid arthritis: A new polymorphism haplotype is associated with increased disease risk.
Clinical immunology (Orlando, Fla.) (2015-05-02)
Jing Sun, Cuiping Liu, Li Gao, Yundi Guo, Yaqin Zhang, Pingping Wu, Juean Jiang, Ruhong Yan, Xueguang Zhang
ZUSAMMENFASSUNG
CD276 (B7-H3) is a costimulatory molecule that plays a potent role in T cell responses, however, the role of B7-H3 in autoimmune diseases has not been elucidated. We analyzed B7-H3 expression in rheumatoid arthritis (RA) for the first time and found B7-H3 was significantly up-regulated on monocytes in RA patients, while the levels of soluble B7-H3 in serum were lower than in controls (P < 0.0001). These differences correlated with clinical and laboratory disease parameters and informatory factor TNF-α. Through in vitro experiments, we demonstrated that B7-H3 promoted TNF-α secretion. In addition, a new polymorphism variant, B7-H3-T-A-C-T, was identified and shown to be associated with the incidence of RA and the decreased release of sB7-H3. These results suggest that B7-H3 may be a promising biomarker associated with the pathogenesis of RA. Notably, the new B7-H3-T-A-C-T polymorphism variant is associated with RA risk and might be associated with the release of soluble B7-H3.