Merck
  • Insulin-like growth factor-1 receptor overexpression is associated with outcome in invasive urothelial carcinoma of urinary bladder: a retrospective study of patients treated using radical cystectomy.
Insulin-like growth factor-1 receptor overexpression is associated with outcome in invasive urothelial carcinoma of urinary bladder: a retrospective study of patients treated using radical cystectomy.
Urology (2014-04-10)
Nilda Gonzalez-Roibon, Jenny J Kim, Sheila F Faraj, Alcides Chaux, Stephania M Bezerra, Enrico Munari, Carla Ellis, Rajni Sharma, Daniel Keizman, Trinity J Bivalacqua, Mark Schoenberg, Mario Eisenberger, Michael Carducci, George J Netto
ZUSAMMENFASSUNG
To assess the insulin-like growth factor-1 receptor (IGF1R) expression in urothelial carcinoma (UC) and its prognostic role in relation to clinicopathologic parameters. A total of 100 cases of invasive UC were evaluated using tissue microarrays. Membranous IGF1R staining was evaluated using immunohistochemistry. A scoring method analogous to that of HER2 expression in breast carcinoma was used, and the highest score was assigned in each tumor. IGF1R was considered overexpressed in cases with score≥1. We found IGF1R overexpression in 62% of invasive UC. IGF1R overexpression was associated with race (P=.04) and pT category (P=.03). Median follow-up was 29 months (range, 0.5-212). Progression rate was 60%, and overall mortality and cancer-specific mortality rates were 69% and 51%, respectively. In invasive UC, IGF1R overexpression was significantly associated with overall mortality and cancer-specific mortality (Mantel Cox P=.0002 and P=.006, respectively). IGF1R overexpression was associated with increased hazard ratios (HRs) for overall mortality (HR=2.63, P=.001) and cancer-specific mortality (HR=2.45, P=.01), independently and after adjusting for clinicopathologic features and treatment modalities. We found IGF1R overexpression in 62% of bladder UC. More importantly, IGF1R overexpression was a significant predictor of overall mortality and cancer-specific mortality, suggesting its potential role as a prognosticator in UC of bladder.