Merck
PHF2, a novel PHD finger gene located on human chromosome 9q22.
Mammalian genome : official journal of the International Mammalian Genome Society (1999-03-02)
K Hasenpusch-Theil, B P Chadwick, T Theil, S K Heath, D G Wilkinson, A M Frischauf
ZUSAMMENFASSUNG
We have isolated and characterized a novel PHD finger gene, PHF2, which maps to human Chromosome (Chr) 9q22 close to D9S196. Its mouse homolog was also characterized and mapped to the syntenic region on mouse Chr 13. The predicted human and mouse proteins are 98% identical and contain a PHD finger domain, eight possible nuclear localization signals, two potential PEST sequences, and a novel conserved hydrophobic domain. Northern analysis shows widespread expression of PHF2 in adult tissues, while in situ hybridization on mouse embryos reveals staining in the neural tube and dorsal root ganglia significantly above a ubiquitous low level expression signal. From its expression pattern and its chromosomal localization, PHF2 is a candidate gene for hereditary sensory neuropathy type I, HSN1.