Merck
  • Complete assignment of the 1H and 13C NMR spectra of antimicrobial 4-arylamino- 3-nitrocoumarin derivatives.
Complete assignment of the 1H and 13C NMR spectra of antimicrobial 4-arylamino- 3-nitrocoumarin derivatives.
Magnetic resonance in chemistry : MRC (2010-09-08)
Vidoslav Dekić, Niko Radulović, Rastko Vukićević, Biljana Dekić, Danielle Skropeta, Radosav Palić
ZUSAMMENFASSUNG
Herein, we describe the synthesis and complete assignment of the (1)H and (13)C NMR chemical shifts of a series of antimicrobial 4-arylamino-3-nitrocoumarin derivatives based on a combination of (1)H and (13)C NMR, (1)H-(1)H-COSY, NOESY, HSQC and HMBC experiments. Conformational effects upon the chemical shifts of the coumarin moiety arising from the anisotropy of the aryl side group are briefly discussed. This study provides the first complete and fully assigned NMR data for this important group of antimicrobial compounds and bridges the gap existing in the literature with regard to NMR structural data for 4-arylamino-3-nitrocoumarins.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Hydroxy-3-nitrocumarin, 98%