Merck
  • Immunohistochemical detection of CD1A antigen in formalin-fixed and paraffin-embedded tissue sections with monoclonal antibody 010.
Immunohistochemical detection of CD1A antigen in formalin-fixed and paraffin-embedded tissue sections with monoclonal antibody 010.
The Journal of pathology (1993-10-01)
L Krenács, L Tiszalvicz, T Krenács, L Boumsell
ZUSAMMENFASSUNG
The immunoreactivity of a CD1a monoclonal antibody (MAb), denoted 010, was investigated by means of the streptavidin-biotin-peroxidase method in formalin-fixed and paraffin-embedded tissues from 47 cases. The samples comprised reactive lymphoid proliferations of skin, tonsil, and lymph node including dermatopathic lymphadenopathy and Langerhans' cell histiocytosis, Hodgkin's and non-Hodgkin's lymphomas, and thymomas. Interdigitating and dermal dendritic cells, veiled cells, Langerhans' cells, and also cortical thymocytes and their neoplastic counterparts displayed immunostaining with MAb 010 in paraffin sections. These results are identical to previous ones reported for other CD1a MAbs in fresh or frozen specimens. The findings suggest that the binding site of 010 is a fixation-resistant epitope of CD1a antigen which has not been previously identified.