Merck
  • Ultrastructural immunohistochemical localization of gastrin, somatostatin and serotonin in endocrine cells of human antral gastric mucosa.
Ultrastructural immunohistochemical localization of gastrin, somatostatin and serotonin in endocrine cells of human antral gastric mucosa.
Acta histochemica (2003-07-02)
Maria Angelova Tzaneva
ZUSAMMENFASSUNG
Five types of endocrine cells are found in the human antral gastric mucosa: gastrin (G) cells, somatostatin (D) cells, enterochromaffin (EC) cells and cells with an unknown secretory product (D1 cells and P cells). The content of secretory granules, gastrin, somatostatin and serotonin, was evaluated using electron microscopic immunohistochemistry and was compared with the granular content in G cells, D cells and EC cells as determined by routine electron microscopy. Semi-quantitative scoring of the granular content was performed on a scale 1-4 (empty-full). The content of gastrin (2.5 +/- 0.2) and somatostatin (3.3 +/- 0.2) in the granules was not different from the granular content in G cells (2.5 +/- 0.3; p > 0.05) and D cells (3.5 +/- 0.2; p > 0.05). Gastrin was also found in G cells in a nongranular form. The content of serotonin in granules (2.8 +/- 0.3) was smaller than the granular content in EC cells (3.7 +/- 0.2; p < 0.05). In intermediate-full and intermediate-empty granules, serotonin was localized in the periphery of granules whereas the granular content in EC cells was localized in an eccentric or central pattern. The granular content of D1 cells and P cells was 3.8 +/- 0.2, and 3.4 +/- 0.2, respectively. It is concluded that gastrin and somatostatin immunostaining in granules of G cells and D cells reflects the granular content in G cells and D cells, respectively, whereas serotonin immunostaining does not agree with the granular content of EC cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Somatostatin (EP130) Rabbit Monoclonal Primary Antibody