Merck
The B29 and mb-1 polypeptides are differentially expressed during human B cell differentiation.
European journal of immunology (1992-10-01)
D Y Mason, C J van Noesel, J L Cordell, W M Comans-Bitter, K Micklem, A G Tse, R A van Lier, J J van Dongen
ZUSAMMENFASSUNG
Surface immunoglobulin on mouse B cells is associated with a heterodimer comprising the products of the mb-1 and B29 genes. Here we report that antibodies raised against a peptide sequence from the intracytoplasmic C terminus of the B29 murine gene product detect the 37-kDa component of the human heterodimer, indicating that this component in man is also encoded by the B29 gene. The immunocytochemical reactivity of these anti-B29 antibodies was compared with those of antibodies to the mb-1 protein. Of 25 cases of acute lymphoblastic leukaemia (ALL), 24 were positive for mb-1 whereas B29 was expressed in only 13 cases. Most of these B29-positive ALL expressed immunoglobulin mu heavy chain in their cytoplasm (pre-B ALL). In lymphoid tissue sections, anti-B29-labeled B cell follicles in a similar fashion to anti-mb-1, with the striking exception that plasma cells were unreactive for B29, but positive for mb-1. These results suggest that the synthesis of B29 begins later in precursor B cells than that of mb-1, and ceases before the terminal plasmacyte phase.