Merck
  • ZBP-89, a Krüppel-like zinc finger protein, inhibits epidermal growth factor induction of the gastrin promoter.
ZBP-89, a Krüppel-like zinc finger protein, inhibits epidermal growth factor induction of the gastrin promoter.
Molecular and cellular biology (1996-12-01)
J L Merchant, G R Iyer, B R Taylor, J R Kitchen, E R Mortensen, Z Wang, R J Flintoft, J B Michel, R Bassel-Duby
ZUSAMMENFASSUNG
We have shown previously that a GC-rich element (GGGGCGGGGTGGGGGG) conferring epidermal growth factor (EGF) responsiveness to the human gastrin promoter binds Sp1 and additional undefined complexes. A rat GH4 cell line expression library was screened by using a multimer of the gastrin EGF response element, and three overlapping cDNA clones were identified. The full-length rat cDNA encoded an 89-kDa zinc finger protein (ZBP-89) that was 89% identical to a 49-kDa human factor, ht(beta), that binds a GTGGG/CACCC element in T-cell receptor promoters. The conservation of amino acids between the zinc fingers indicates that ZBP-89 is a member of the C2H2 zinc finger family subclass typified by the Drosophila Krüppel protein. ZBP-89 is ubiquitously expressed in normal adult tissues. It binds specifically to the gastrin EGF response element and inhibits EGF induction of the gastrin promoter. Collectively, these results demonstrate that ZBP-89 functions as a repressor of basal and inducible expression of the gastrin gene.