Merck
  • Identification of a novel gene, DAM1, amplified at chromosome 1p13.3-21 region in human breast cancer cell lines.
Identification of a novel gene, DAM1, amplified at chromosome 1p13.3-21 region in human breast cancer cell lines.
Cancer letters (1999-07-14)
K Nagasaki, N Maass, T Manabe, H Hanzawa, T Tsukada, K Kikuchi, K Yamaguchi
ZUSAMMENFASSUNG
Screening for differentially expressed genes in cancer cell lines by the RNA differential display (DD) technique identified a novel mRNA that encodes a 26-kDa protein and is up-regulated in MCF-7 and BT-20 breast cancer cells. The predicted amino acid sequence showed 38% homology to Caenorhabditis elegans T12A2.7 protein, indicating that this is a possible human homologue for C. elegans T12A2.7 protein. The mRNA, designated DAM1 (DNA amplified in mammary carcinoma), was mapped at the 1p13.3-21 region, which is frequently altered in human breast cancer. By Southern blot analysis, we confirmed that the up-regulation of this novel gene is associated with gene amplification in MCF-7 (10-fold) and BT-20 (5-fold) cells. Our findings suggest that the DAM1 gene is a novel gene up-regulated by amplification in human breast cancer cell lines.