- Molecular interactions of coreceptor CD8 and MHC class I: the molecular basis for functional coordination with the T-cell receptor.
Molecular interactions of coreceptor CD8 and MHC class I: the molecular basis for functional coordination with the T-cell receptor.
Immunology today (2000-12-15)
G F Gao, B K Jakobsen
PMID11114424
ZUSAMMENFASSUNG
In recent years, substantial progress has been made towards understanding the molecular basis for CD8 binding to class I MHC and the coreceptor's role in cytotoxic T-cell activation. Here, we review the structural, mechanistic and functional studies that point to a model of coordination of T-cell receptor and CD8 signaling that might provide the key to cytotoxic T-cell activation.