- Celiprolol-induced lupus-like syndrome.
Celiprolol-induced lupus-like syndrome.
Acta cardiologica (2007-01-09)
Jean-Christophe Charniot, Thierry Genereau, Luc Mouthon
PMID17205925
ZUSAMMENFASSUNG
Systemic lupus erythematosus can be idiopathic or drug-induced. Although a number of beta-blockers have been reported to induce a lupus-like syndrome, to the best of our knowledge, no such case has been described following celiprolol therapy. We diagnosed a lupus-like syndrome in a 67-year-old female patient who developed febrile polyarthritis, percarditis, antinuclear and anti-histone antibodies after taking celiprolol for 2 years. Despite drug withdrawal, prolonged corticotherapy was needed to obtain clinical and biological remission.