Merck
Mechanistic insights into the stepwise Diels-Alder reaction of 4,6-dinitrobenzofuroxan.
Organic letters (2011-12-01)
Mats Linder, Adam Johannes Johansson, Tore Brinck
ZUSAMMENFASSUNG
The stepwise Diels-Alder reaction between 1-trimethylsiloxy-1,3-butadiene and 4,6-dinitrobenzofuroxan is explored using state-of-the-art computational methods. The results support a stepwise mechanism via a persistent intermediate, however, not the one previously reported (Lakhdar et al., Chem. Eur. J.2007, 16, 5681) but a heterocyclic adduct. The novel DFT functional M062X and the SCS-MP2 method were essential to reproduce a reasonable potential energy surface for this challenging system.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1-(Trimethylsiloxy)-1,3-butadien, 98%