Merck
Inorganic and total arsenic contents in rice-based foods for children with celiac disease.
Journal of food science (2013-12-10)
Sandra Munera-Picazo, Amanda Ramírez-Gandolfo, Francisco Burló, Angel Antonio Carbonell-Barrachina
ZUSAMMENFASSUNG
Celiac disease is an autoimmune disease that affects the villi of the small intestine causing abdominal pain, gas, diarrhea, or bad absorption due to gluten intolerance. The only treatment for this disease consists of a lifelong gluten free diet; this is, celiac people cannot consume products containing gluten, such as wheat, barley, and rye, but they can use rice and corn. Thus, rice flour is mainly used for the manufacturing of the basic products of this population. Unfortunately, rice can contain high contents of total (t-As) and inorganic (i-As) arsenic. The current study demonstrated that products for celiac children with a high percentage of rice contained high concentrations of arsenic (256 and 128 μg kg⁻¹). The daily intake of i-As ranged from 0.61 to 0.78 μg kg⁻¹ body weight (bw) in children up to 5 y of age; these values were below the maximum value established by the EFSA Panel (8.0 μg kg⁻¹ bw per day), but it should be considered typical of populations with a high exposure to this pollutant. Finally, legislation is needed to improve the labeling of these special rice-based foods for celiac children; label should include information about percentage, geographical origin, and cultivar of the used rice.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Reismehl (Gesamt-As und As-Verbindungen), ERM®, certified reference material