Merck
Tbx19, a tissue-selective regulator of POMC gene expression.
Proceedings of the National Academy of Sciences of the United States of America (2001-07-12)
J Liu, C Lin, A Gleiberman, K A Ohgi, T Herman, H P Huang, M J Tsai, M G Rosenfeld
ZUSAMMENFASSUNG
Pituitary cell types arise in a temporally and spatially specific fashion, in response to combinatorial actions of transcription factors induced by transient signaling gradients. The critical transcriptional determinants of the two pituitary cell types that express the pro-opiomelanocortin (POMC) gene, the anterior lobe corticotropes, producing adrenocorticotropin, and the intermediate lobe melanotropes, producing melanocyte-stimulating hormone (MSH alpha), have remained unknown. Here, we report that a member of the T-box gene family, Tbx19, which is expressed only in the rostral ventral diencephalon and pituitary gland, commencing on e11.5, marks pituitary cells that will subsequently express the POMC gene and is capable of altering progression of ventral cell types and inducing adrenocorticotropin in rostral tip cells. It is suggested that Tbx19, depending on the presence of synergizing transcription factors, can activate POMC gene expression and repress the alpha glycoprotein subunit and thyroid-stimulating hormone beta promoters.