Merck
[Effect of thiochrome capronate on lipid metabolism in rats].
Ukrainskii biokhimicheskii zhurnal (1978) (1986-09-01)
F S Larin, V U Buko, T I Zimatkina, D O Oparin, Iu M Ostrovskiĭ
ZUSAMMENFASSUNG
Thiochrome caproate modified by the oxyethyl radical of the thiochrome derivative was studied for its effect on different indices of lipid metabolism in the liver and blood of albino rats. It was shown that when animals were fed on a standard laboratory diet, thiochrome caproate changed the amount of total and free fatty acids in the studied tissues and the fatty acid composition in the liver to a greater extent than thiochrome and hydroxythiamine.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Thiochrom, analytical standard