Merck
  • A sulfhydryl-reactive ruthenium (II) complex and its conjugation to protein G as a universal reagent for fluorescent immunoassays.
A sulfhydryl-reactive ruthenium (II) complex and its conjugation to protein G as a universal reagent for fluorescent immunoassays.
PloS one (2012-05-09)
Jing-Tang Lin, Po-Chung Chen, Thirumani Venkatshwar Goud, Bor-Rong Huang, Tzu-Chau Lin, Jean-François Biellmann, Chien-Sheng Chen
ZUSAMMENFASSUNG
To develop a fluorescent ruthenium complex for biosensing, we synthesized a novel sulfhydryl-reactive compound, 4-bromophenanthroline bis-2,2'-dipyridine Ruthenium bis (hexafluorophosphate). The synthesized Ru(II) complex was crosslinked with thiol-modified protein G to form a universal reagent for fluorescent immunoassays. The resulting Ru(II)-protein G conjugates were identified by sodium dodecyl sulfate-polyacrylamide gel electrophoresis (SDS-PAGE). The emission peak wavelength of the Ru(II)-protein G conjugate was 602 nm at the excitation of 452 nm which is similar to the spectra of the Ru(II) complex, indicating that Ru(II)-protein G conjugates still remain the same fluorescence after conjugation. To test the usefulness of the conjugate for biosensing, immunoglobulin G (IgG) binding assay was conducted. The result showed that Ru(II)-protein G conjugates were capable of binding IgG and the more cross-linkers to modify protein G, the higher conjugation efficiency. To demonstrate the feasibility of Ru(II)-protein G conjugates for fluorescent immunoassays, the detection of recombinant histidine-tagged protein using the conjugates and anti-histidine antibody was developed. The results showed that the histidine-tagged protein was successfully detected with dose-response, indicating that Ru(II)-protein G conjugate is a useful universal fluorescent reagent for quantitative immunoassays.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
S-Acetylthioglykolsäure-N-hydroxysuccinimid-Ester, ≥95% (TLC), powder