Merck
  • Plasmodium vinckei: infectivity of arteether-sensitive and arteether-resistant parasites in different strains of mice.
Plasmodium vinckei: infectivity of arteether-sensitive and arteether-resistant parasites in different strains of mice.
Parasitology research (2011-04-12)
Ramesh Chandra, Santosh Kumar, S K Puri
ZUSAMMENFASSUNG
Malaria is one of the most lethal parasitic infections in the world. The lethality of the parasite depends on the rate of multiplication of the parasite within host erythrocytes. Different strains of the malaria parasite often respond in a different way to the same strain of mice or vice versa. In the present study, we investigated the course of infection of the arteether-sensitive and arteether-resistant Plasmodium vinckei parasites in Swiss albino AKR (inbred) and AJ (outbred) mice. The higher parasite burden and mortality were observed in the sensitive parasite-infected mice, whereas the infection with the resistant parasite was non-lethal. Resistant parasite-infected mice developed a moderate level of parasitemia that decreased gradually throughout the infection. The microscopic examination suggests that the resistant parasite invades reticulocytes more efficiently than normocytes, regardless of the mouse strain examined. Since the reticulocytes are rare in blood circulation, it limits the increase in parasite proliferations, while arteether-sensitive parasites can invade both mature normocytes and reticulocytes, resulting in the mortality of the mice. However, treatment with phenylhydrazine in Swiss mice results in reticulocytosis, which transforms the non-lethal resistant parasites to produce lethal infections. Our findings demonstrate that the characteristic response during infections with the arteether-resistant strain is dependent on the availability of reticulocytes in peripheral blood circulation. We can use this model for identifying the interaction between host and artemisinin derivative-resistant parasites.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phenylhydrazin -hydrochlorid, ≥99%
Sigma-Aldrich
Phenylhydrazin, 97%