Merck
  • Pomegranate (Punica granatum) purified polyphenol extract inhibits influenza virus and has a synergistic effect with oseltamivir.
Pomegranate (Punica granatum) purified polyphenol extract inhibits influenza virus and has a synergistic effect with oseltamivir.
Phytomedicine : international journal of phytotherapy and phytopharmacology (2009-07-10)
Mehran Haidari, Muzammil Ali, Samuel Ward Casscells, Mohammad Madjid
ZUSAMMENFASSUNG
Influenza epidemics cause numerous deaths and millions of hospitalizations each year. Because of the alarming emergence of resistance to anti-influenza drugs, there is a need to identify new naturally occurring antiviral molecules. We tested the hypothesis that pomegranate polyphenol extract (PPE) has anti-influenza properties. Using real time PCR, plaque assay, and TCID 50% hemagglutination assay, we have shown that PPE suppresses replication of influenza A virus in MDCK cells. PPE inhibits agglutination of chicken red blood cells (cRBC) by influenza virus and is virucidal. The single-cycle growth conditions indicated that independent of the virucidal effect PPE also inhibits viral RNA replication. PPE did not alter virus ribonucleoprotein (RNP) entry into nucleus or translocation of virus RNP from nucleus to cytoplasm in MDCK cells. We evaluated four major Polyphenols in PPE (ellagic acid, caffeic acid, luteolin, and punicalagin) and demonstrated that punicalagin is the effective, anti-influenza component of PPE. Punicalagin blocked replication of the virus RNA, inhibited agglutination of chicken RBC's by the virus and had virucidal effects. Furthermore, the combination of PPE and oseltamivir synergistically increased the anti-influenza effect of oseltamivir. In conclusion, PPE inhibited the replication of human influenza A/Hong Kong (H3N2) in vitro. Pomegranate extracts should be further studied for therapeutic and prophylactic potential especially for influenza epidemics and pandemics.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Punicalagin, ≥98% (HPLC), from pomegranate
Supelco
Punicalagin, analytical standard