Merck
Derivatives of pyrazinecarboxylic acid: 1H, 13C and 15N NMR spectroscopic investigations.
Magnetic resonance in chemistry : MRC (2009-04-30)
Wolfgang Holzer, Gernot A Eller, Barbara Datterl, Daniela Habicht
ZUSAMMENFASSUNG
NMR spectroscopic studies are undertaken with derivatives of 2-pyrazinecarboxylic acid. Complete and unambiguous assignment of chemical shifts ((1)H, (13)C, (15)N) and coupling constants ((1)H,(1)H; (13)C,(1)H; (15)N,(1)H) is achieved by combined application of various 1D and 2D NMR spectroscopic techniques. Unequivocal mapping of (13)C,(1)H spin coupling constants is accomplished by 2D (delta,J) long-range INEPT spectra with selective excitation. Phenomena such as the tautomerism of 3-hydroxy-2-pyrazinecarboxylic acid are discussed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyrazincarbonsäure, 99%