Merck
Cranial osteomyelitis: a late complication of a dental infection.
The British journal of oral & maxillofacial surgery (2008-05-20)
James R Adams, Douglas G Bryant
ZUSAMMENFASSUNG
Dental infections are associated with a range of serious complications. The orofacial region provides potential spaces in the tissue that infections of dental origin can occupy. We describe the subtemporal extension of a dental infection, the late development of cranial osteomyelitis, and its surgical management.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cefradin, ≥90.0% (Cephradine, HPLC)