Merck
Fabrication of amino silane-coated microchip for DNA extraction from whole blood.
Journal of biotechnology (2005-01-25)
Takahito Nakagawa, Tsuyoshi Tanaka, Daisuke Niwa, Tetsuya Osaka, Haruko Takeyama, Tadashi Matsunaga
ZUSAMMENFASSUNG
A simple microchip device for DNA extraction was constructed based on electrostatic interactions between surface amine groups and DNA. Microchannel was fabricated on silicon wafer by photolithography and coated with 3-aminopropyltriethoxysilane (APTES) or 3-[2-(2-aminoethylamino)-ethylamino]-propyltrimethoxysilane (AEEA) to introduce amine groups on the surface. Determination of the number of surface amine groups and optimization of DNA capture condition were demonstrated to characterize the microchip. Capacities of capturing DNA were approximately 97 ng/cm2 in APTES and 194 ng/cm2 in AEEA modified microchips, respectively. The amount of DNA captured in the microchip increased depending on surface amine density. Furthermore, DNA extraction using amine-coated microchip from whole blood was examined. Quantification of DNA and proteins in washing or eluting fraction indicates that proteins were removed at washing steps and only DNA was effectively eluted by changing alkalinity of buffer from pH 7.5 to 10.6. The amount of DNA extracted from whole blood was approximately 10 ng and its recovery ratio was 27-40%. Performance of PCR for the eluted fraction indicates that DNA extracted from whole blood was well purified using amine-coated microchip.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-[2-(2-Aminoethylamino)-ethylamino]-propyltrimethoxysilan, technical grade