Merck
  • Sphingosylphosphorylcholine stimulates CCL2 production from human umbilical vein endothelial cells.
Sphingosylphosphorylcholine stimulates CCL2 production from human umbilical vein endothelial cells.
Journal of immunology (Baltimore, Md. : 1950) (2011-03-04)
Ha Young Lee, Sun Young Lee, Sang Doo Kim, Jae Woong Shim, Hak Jung Kim, Young Su Jung, Jae Young Kwon, Suk-Hwan Baek, Junho Chung, Yoe-Sik Bae
ZUSAMMENFASSUNG
Sphingosylphosphorylcholine (SPC) is a component of high-density lipoprotein particles. We investigated the functional role of SPC in HUVECs. SPC stimulation induced production of the CCL2 chemokine in a PTX-sensitive G-protein-dependent manner. SPC treatment caused the activation of NF-κB and AP-1, which are essential for SPC-induced CCL2 production, and induced the activation of three MAPKs, ERK, p38 MAPK, and JNK. Inhibition of p38 MAPK or JNK by specific inhibitors caused a dramatic decrease in SPC-induced CCL2 production. The Jak/STAT3 pathway was also activated upon SPC stimulation of HUVECs. Pretreatment with a Jak inhibitor blocked not only SPC-induced p38 MAPK and JNK activation, but also NF-κB and AP-1 activation. Our results suggest that SPC stimulates HUVECs, resulting in Jak/STAT3-, NF-κB-, and AP-1-mediated CCL2 production. We also observed that SPC stimulated expression of the adhesion molecule ICAM-1 in HUVECs. Our results suggest that SPC may contribute to atherosclerosis; therefore, SPC and its unidentified target receptor offer a starting point for the development of a treatment for atherosclerosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Sphingosylphosphorylcholin, ≥98%, powder