Merck
  • Biologic effects of heregulin/neu differentiation factor on normal and malignant human breast and ovarian epithelial cells.
Biologic effects of heregulin/neu differentiation factor on normal and malignant human breast and ovarian epithelial cells.
Oncogene (1999-11-11)
Z Aguilar, R W Akita, R S Finn, B L Ramos, M D Pegram, F F Kabbinavar, R J Pietras, P Pisacane, M X Sliwkowski, D J Slamon
ZUSAMMENFASSUNG
The heregulins are a family of ligands with ability to induce phosphorylation of the p185HER-2/neu receptor. Various investigators have reported a variety of responses of mouse and human breast and ovarian cells to this family of ligands including growth stimulation, growth inhibition, apoptosis and induction of differentiation in cells expressing the HER-2/neu receptor. Some of the disparity in the literature has been attributed to variations in the cell lines studied, ligand dose applied, methodologies utilized or model system evaluated (i.e. in vitro or in vivo). To evaluate the effects of heregulin on normal and malignant human breast and ovarian epithelial cells expressing known levels of the HER-2/neu receptor, this report presents the use of several different assays, performed both in vitro and in vivo, in vitro proliferation assays, direct cell counts, clonogenicity under anchorage-dependent and anchorage-independent conditions, as well as the in vivo effects of heregulin on human cells growing in nude mice to address heregulin activity. Using a total of five different biologic assays in nine different cell lines, across two different epithelia and over a one log heregulin dose range, we obtained results that clearly indicate a growth-stimulatory role for this ligand in human breast and ovarian epithelial cells. We find no evidence that heregulin has any growth-inhibitory effects in human epithelial cells. We also quantitated the amount of each member of the type I receptor tyrosine kinase family (RTK I, i.e. HER-1, HER-2, HER-3 and HER-4) in the cell lines employed and correlated this to their respective heregulin responses. These data demonstrate that HER-2/neu overexpression itself affects the expression of other RTK I members and that cells expressing the highest levels of HER-2/neu have the greatest response to HRG.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Heregulin-α, EGF Domain human, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, ≥97% (SDS-PAGE)