Merck
Lysine Methyltransferase 2D Regulates Immune Response and Metastasis in Head and Neck Cancer.
Anticancer research (2024-07-27)
Jianchun Wu, Crystal Chun, Angelica M Lagunas, David L Crowe
ZUSAMMENFASSUNG
The most frequently altered epigenetic modifier in head and neck squamous carcinoma (HNSC) is the histone methyltransferase KMT2D. KMT2D catalyzes methylation of histone H3K4 resulting in open chromatin and the activation of target genes. Tumor-associated macrophages (TAMs) promote cancer growth by causing T lymphocyte exhaustion. C-C motif chemokine ligand 2 (CCL2) is a potent TAM chemotactic factor. In HNSC, TAMs have been associated with unfavorable patient outcomes and metastasis. The aim of this study was to determine the role of KMT2D in HNSC using genetically engineered in vivo models. KMT2D protein expression was correlated with lymph node metastasis in human HNSC using immunohistochemistry. Genetically engineered KMT2D and CCL2 knockout models of HNSC were created in vivo. HNSC was characterized using qRT-PCR, histopathology, and immunohistochemistry/immunofluorescence microscopy. We also analyzed the effects of KMT2D expression on the proliferation and migration of human HNSC lines. The regulation of the CCL2 gene by KMT2D was characterized using chromatin immunoprecipitation-sequencing assay of transposase accessible chromatin-sequencing, and chromatin conformation capture-sequencing. Human HNSC cases with high KMT2D expression exhibited significantly increased lymph node metastasis. Reduced KMT2D expression in our genetically engineered model correlated with reduced lymph node metastasis, longer latency, and slow tumor growth. CCL2 expression was decreased in KMT2D deficient HNSC, which correlated with a reduced TAM gene expression signature. Genomic experiments demonstrated that KMT2D directly targeted the CCL2 gene. A new genetically engineered in vivo model of CCL2-null HNSC was created, recapitulating the KMT2D deficient phenotype and showing a decreased T lymphocyte exhaustion signature. KMT2D regulates CCL2-mediated immune response and metastasis in HNSC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-KMT2D antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution