Merck
  • Jack bean (Canavalia ensiformis) urease. Probing acid-base groups of the active site by pH variation.
Jack bean (Canavalia ensiformis) urease. Probing acid-base groups of the active site by pH variation.
Plant physiology and biochemistry : PPB (2005-07-19)
Barbara Krajewska, Stefano Ciurli
ZUSAMMENFASSUNG
A pH-variation study of jack bean (Canavalia ensiformis) urease steady-state kinetic parameters and of the inhibition constant of boric acid, a urease competitive inhibitor, was performed using both noninhibitory organic (MES, HEPES and CHES) and inhibitory inorganic (phosphate) buffers, in an effort to elucidate the functions exercised in the catalysis by the ionizable groups of the enzyme active site. The results obtained are consistent with the requirement for three groups utilized by urease with pK(a)s equal to 5.3+/-0.2, 6.6+/-0.2 and 9.1+/-0.4. Based on the appearance of the ionization step with pK(a)=5.3 in v(max)-pH, K(M)-pH and K(i)-pH profiles, we assigned this group as participating both in the substrate binding and catalytic reaction. As shown by its presence in v(max)-pH and K(M)-pH curves, the obvious role of the group with pK(a)=9.1 is the participation in the catalytic reaction. One function of the group featuring pK(a)=6.6, which was derived from a two-maxima v(max)-pH profile obtained upon increasing phosphate buffer concentration, an effect the first time observed for urease-phosphate systems, is the substrate binding, another possible function being modulation of the active site structure controlled by the ionic strength. It is also possible that the pK(a)=6.6 is a merger of two pK(a)s close in value. The study establishes that regular bell-shaped activity-pH profiles, commonly reported for urease, entail more complex pH-dependent behavior of the urease active site ionizable groups, which could be experimentally derived using species interacting with the enzyme, in addition to changing solution pH and ionic strength.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Urease aus Canavalia ensiformis (Jack bean), Type C-3, powder, ≥600,000 units/g solid